Please login to your online account to display your discounted pricing

anti-TACI/TNFRSF13B/CVID, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056516

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056516

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Gene ID (Entrez) 23495
Conjugate Unlabeled
Regulatory Status RUO
Immunogen This rabbit polyclonal antibody was developed against a synthetic peptide corresponding to amino acids 116-132 of human TACI. Human and mouse TACI amino acid sequences are 65% conserved in the immunogen.
Cross Reactivity Human
Cross Reactivity Mouse
Formulation PBS containing 0.2% gelatin
Monoclonal or Polyclonal Polyclonal
Format Purified
Gene Accession No. NP_036584
Primary or Secondary Primary
Quantity 0.1mg
Applications Western Blotting
Antigen TACI/TNFRSF13B/CVID
Host Species Rabbit