missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TACI/TNFRSF13B/CVID, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen This rabbit polyclonal antibody was developed against a synthetic peptide corresponding to amino acids 116-132 of human TACI. Human and mouse TACI amino acid sequences are 65% conserved in the immunogen.
Antigen TACI/TNFRSF13B/CVID
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056516 Novus Biologicals Canada
NB10056516
0.1mg Each for $259.02
NB10056516T Novus Biologicals Canada
NB10056516SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
TACI/TNFRSF13B/CVID
Purified
RUO
NP_036584
PBS containing 0.2% gelatin
This rabbit polyclonal antibody was developed against a synthetic peptide corresponding to amino acids 116-132 of human TACI. Human and mouse TACI amino acid sequences are 65% conserved in the immunogen.
Polyclonal
Primary
Western Blot
23495
0.5mg/mL