Please login to your online account to display your discounted pricing

anti-TA4/TAAR6 Polyclonal, Thermo Scientific™

TA4/TAAR6 Polyclonal antibody PA5-34242 from Thermo Scientific™ specifically detects TA4/TAAR6 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

$506.05 - $506.05

Specifications

Antigen TA4/TAAR6
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 14 amino acid peptide from C-terminus of human TAAR6.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIPA534241

 
pierce biotechnology inc
PA534241
Unlabeled 50μg Each for $506.05

PIPA534242

 
pierce biotechnology inc
PA534242
Unlabeled 50μg Each for $506.05
Description & Specifications

Specifications

Antigen TA4/TAAR6
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 14 amino acid peptide from C-terminus of human TAAR6.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C

TA4/TAAR6 Polyclonal antibody PA5-34242 from Thermo Scientific™ specifically detects TA4/TAAR6 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

L-glutamate is the major excitatory neurotransmitter in the central nervous system and activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors, that have been divided into 3 groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties. Group I includes GRM1 and GRM5 and these receptors have been shown to activate phospholipase C. Group II includes GRM2 and GRM3 while Group III includes GRM4, GRM6, GRM7 and GRM8. Group II and III receptors are linked to the inhibition of the cyclic AMP cascade but differ in their agonist selectivities. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 14 amino acid peptide from C-terminus of human TAAR6.
Formulation: 50μg purified Antibody (1mg/mL) in PBS, with < 0.1% sodium azide;
Synonyms: TAAR6, TA4, TaR-4, TAR4, TAR6, TaR-6, TRAR4, Trace amine receptor 4, Trace amine receptor 6, RP11-295F4.3
Uniprot Accession: Q96RI8
Gene Symbol: TAAR6
Entrez GeneID: 319100
PA5-33832 detects GRM8/MGLUR8 in Human samples. PA5-33832 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA5-33832 The immunogen is Synthetic 19 amino acid peptide from C-terminus of human GRM8/MGLUR8..