Please login to your online account to display your discounted pricing

anti-TA3/TAAR9 Polyclonal, Thermo Scientific™


TA3/TAAR9 Polyclonal antibody PA5-34245 from Thermo Scientific™ specifically detects TA3/TAAR9 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

Manufacturer: pierce biotechnology inc  PA534245

 View more versions of this product

Catalog No. PIPA534245

  • / Each
 

Description & Specifications

Specifications

Formulation 50μg purified antibody (1mg/mL) in PBS and < 0.1% sodium azide
Gene ID (Entrez) 134860
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C
Applications Immunohistochemistry (Paraffin)
Gene Symbols TAAR9
Host Species Rabbit
Gene Accession No. Q96RI9
Gene Alias TAAR9, SNORF1 receptor, TaR-3, TaR-9, TAR3, TAR9, Trace amine receptor 9, TRAR3, TA3, Trace amine receptor 3
Species Reactivity Human
Antigen TA3/TAAR9
Format Purified
Immunogen Synthetic 12 amino acid peptide from 2nd extracellular domain of human TAAR9.
Conjugate Unlabeled
Cross Reactivity Human
Isotype IgG
Quantity 50μg
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal

TA3/TAAR9 Polyclonal antibody PA5-34245 from Thermo Scientific™ specifically detects TA3/TAAR9 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

TAAR9 is a member of a large family of rhodopsin G protein-coupled receptors (GPCRs, or GPRs). GPCRs contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 12 amino acid peptide from 2nd extracellular domain of human TAAR9.
Formulation: 50 ug purified antibody (1mg/mL) in PBS and < 0.1% sodium azide
Synonyms: TAAR9, SNORF1 receptor, TaR-3, TaR-9, TAR3, TAR9, Trace amine receptor 9, TRAR3, TA3, Trace amine receptor 3
Uniprot Accession: Q96RI9
Entrez GeneID: 134860
Gene Symbol: TAAR9
Percent identity with other species by BLAST analysis: Human, Galago, Marmoset, Mouse, Rat, Hamster, Pig (100%) Bovine (92%) Horse (83%).