Please login to your online account to display your discounted pricing

anti-TA3/TAAR9 Polyclonal, Thermo Scientific™

TA3/TAAR9 Polyclonal antibody PA5-34245 from Thermo Scientific™ specifically detects TA3/TAAR9 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

$509.25 - $509.25

Specifications

Antigen TA3/TAAR9
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human TAAR9.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIPA534245

 
pierce biotechnology inc
PA534245
Unlabeled 50μg Each for $509.25

PIPA534246

 
pierce biotechnology inc
PA534246
Unlabeled 50μg Each for $509.25
Description & Specifications

Specifications

Antigen TA3/TAAR9
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human TAAR9.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C

TA3/TAAR9 Polyclonal antibody PA5-34245 from Thermo Scientific™ specifically detects TA3/TAAR9 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

GPR62 has been reported to be expressed in several brain regions, including basal forebrain, frontal cortex, caudate, putamen, thalamus, and hippocampus. ESTs have been isolated from anaplastic oligodendroglioma and ovarian thecoma fibroma libraries. Mouse and rat brain libraries have also been used to isolate ESTs.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 12 amino acid peptide from 2nd extracellular domain of human TAAR9.
Formulation: 50μg purified Antibody (1mg/mL) in PBS, with < 0.1% sodium azide;
Synonyms: TAAR9, SNORF1 receptor, TaR-3, TaR-9, TAR3, TAR9, Trace amine receptor 9, TRAR3, TA3, Trace amine receptor 3
Uniprot Accession: Q96RI9
Gene Symbol: TAAR9
Entrez GeneID: 134860
PA5-33745 detects GPR62 in Human samples. PA5-33745 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA5-33745 The immunogen is Synthetic 16 amino acid peptide from N-terminal extracellular domain of human GPR62..