Please login to your online account to display your discounted pricing

anti-Syncollin, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human syncollin was used as the immunogen.
Antigen Syncollin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224491

 
novus biologicals canada
NBP224491
Unlabeled 0.1mg Each for $259.02

NBP224491SS

 
novus biologicals canada
NBP224491SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human syncollin was used as the immunogen.
Antigen Syncollin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and 85% homologous in bovine.
Gene Accession No. Q0VAF6
Gene ID (Entrez) 342898
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL