Please login to your online account to display your discounted pricing

anti-Syncollin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224491SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224491SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. Q0VAF6
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 342898
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and 85% homologous in bovine.
Format Affinity Purified
Host Species Rabbit
Applications Western Blotting
Immunogen A portion of amino acids 50-100 of human syncollin was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Antigen Syncollin