Please login to your online account to display your discounted pricing

anti-Syncollin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224491

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224491

  • / Each
 

Description & Specifications

Specifications

Antigen Syncollin
Format Affinity Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. Q0VAF6
Host Species Rabbit
Applications Western Blotting
Immunogen A portion of amino acids 50-100 of human syncollin was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 342898
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and 85% homologous in bovine.
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human