Please login to your online account to display your discounted pricing

anti-Synaptophysin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300653SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300653SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 6855
Concentration 1.0mg/mL
Test Specificity The does not cross-react other synaptic vesicle proteins such as synaptotagmin, GAP45, and SNAP25 protiens.
Gene Accession No. P08247
Conjugate Unlabeled
Primary or Secondary Primary
Regulatory Status RUO
Format Affinity Purified
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Immunogen Synthetic peptide from the N-terminal region of human Synaptophysin
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.025mg
Host Species Rabbit
Antigen Synaptophysin