Please login to your online account to display your discounted pricing

anti-Synaptophysin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300653

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300653

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P08247
Gene ID (Entrez) 6855
Test Specificity The does not cross-react other synaptic vesicle proteins such as synaptotagmin, GAP45, and SNAP25 protiens.
Concentration 1.0mg/mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Synthetic peptide from the N-terminal region of human Synaptophysin
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Antigen Synaptophysin
Format Affinity Purified
Host Species Rabbit
Quantity 0.1mg