Please login to your online account to display your discounted pricing

anti-SWAP70, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human SWAP-70 was used as the immunogen.
Antigen SWAP70
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056430T

 
novus biologicals canada
NB10056430SS
Unlabeled 0.025mg Each for $139.00

NB10056430

 
novus biologicals canada
NB10056430
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human SWAP-70 was used as the immunogen.
Antigen SWAP70
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Reacts with Human. Predicted to react with Bovine, Chimpanzee, Dog and Mouse.
Gene Accession No. Q9UH65
Gene ID (Entrez) 23075
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL