Please login to your online account to display your discounted pricing

anti-Survivin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056168SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056168T

  • / Each
 

Description & Specifications

Specifications

Test Specificity Survivin . Cross reacts with Gerbil, Human, Mouse and Rat.
Conjugate Unlabeled
Gene ID (Entrez) 332
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Format Antiserum
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Immunogen Full-length recombinant human survivin protein was used as immunogen.
Antigen Survivin
Quantity 0.025mL
Regulatory Status RUO