Please login to your online account to display your discounted pricing

anti-Survivin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056168

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056168

  • / Each
 

Description & Specifications

Specifications

Antigen Survivin
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Antisera
Host Species Rabbit
Immunogen Full-length recombinant human survivin protein was used as immunogen.
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 332
Test Specificity Survivin . Cross reacts with Gerbil, Human, Mouse and Rat.
Target Species Human
Target Species Murine
Target Species Rat