Please login to your online account to display your discounted pricing

anti-Survivin, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 50-150 of human Survivin was used as the immunogen.
Antigen Survivin
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224395

 
novus biologicals canada
NBP224395
Unlabeled 0.1mg Each for $255.80

NBP224395SS

 
novus biologicals canada
NBP224395SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 50-150 of human Survivin was used as the immunogen.
Antigen Survivin
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and 72% homologous in mouse and rat.
Gene ID (Entrez) 332
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL