Please login to your online account to display your discounted pricing

anti-Survivin, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen Full-length recombinant human survivin protein was used as immunogen.
Antigen Survivin
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056167T

 
novus biologicals canada
NB10056167SS
Unlabeled 0.025mL Each for $144.63

NB10056167

 
novus biologicals canada
NB10056167
Unlabeled 0.05mL Each for $222.95
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Full-length recombinant human survivin protein was used as immunogen.
Antigen Survivin
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Survivin .
Gene Accession No. O15392
Gene ID (Entrez) 332