Please login to your online account to display your discounted pricing

anti-Survivin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224395SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224395SS

  • / Each
 

Description & Specifications

Specifications

Antigen Survivin
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Host Species Rabbit
Immunogen A portion of amino acids 50-150 of human Survivin was used as the immunogen.
Isotype IgG
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 332
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and 72% homologous in mouse and rat.
Target Species Human