Please login to your online account to display your discounted pricing

anti-Survivin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224395

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224395

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Conjugate Unlabeled
Antigen Survivin
Format Purified
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Host Species Rabbit
Applications Western Blotting
Immunogen A portion of amino acids 50-150 of human Survivin was used as the immunogen.
Isotype IgG
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 332
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and 72% homologous in mouse and rat.
Cross Reactivity Human