Please login to your online account to display your discounted pricing

anti-Survivin (p Ser20), Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide surrounding the phosphorylated serine 20 of the human Survivin protein. [Swiss-Prot No.O15392]
Antigen Survivin (p Ser20)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11092717

 
novus biologicals canada
NB11092717
Unlabeled 0.1mL Each for $309.05

NB11092717T

 
novus biologicals canada
NB11092717SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide surrounding the phosphorylated serine 20 of the human Survivin protein. [Swiss-Prot No.O15392]
Antigen Survivin (p Ser20)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Gene Accession No. O15392
Gene ID (Entrez) 332
Formulation PBS, 30% glycerol
Concentration 1.25mg/mL