Please login to your online account to display your discounted pricing

anti-SUR1, Polyclonal, Novus Biologicals™

$450.00 - $450.00

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the Middle Region of the target protein.
Antigen SUR1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

19600002

 
novus biologicals canada
19600002
Unlabeled 0.05mg Each for $450.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the Middle Region of the target protein.
Antigen SUR1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Test Specificity This product is specific for Human SUR1.
Gene Accession No. NM_000352
Gene ID (Entrez) 6833
Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0