missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-SUMO3, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-95 of human SUMO3 (SMT3) was used as the immunogen for this antibody.
Antigen SUMO3
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224651 Novus Biologicals Canada
NBP224651
0.1mg Each for $259.02
NBP224651SS Novus Biologicals Canada
NBP224651SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
SUMO3
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, monkey and chimp, 93% homologous in cow, dog and horse, 87% homologous in rat and 75% homologous in mouse.
6612
0.5mg/mL
A portion of amino acids 50-95 of human SUMO3 (SMT3) was used as the immunogen for this antibody.
Polyclonal
Primary
Western Blot
P61957
100μg antibody in 200μL PBS containing 0.05% BSA