Please login to your online account to display your discounted pricing

anti-SUMO3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224651SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224651SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey and chimp, 93% homologous in cow, dog and horse, 87% homologous in rat and 75% homologous in mouse.
Gene Accession No. P61957
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Primary or Secondary Primary
Gene ID (Entrez) 6612
Regulatory Status RUO
Antigen SUMO3
Host Species Rabbit
Applications Western Blotting
Quantity 0.025mg
Immunogen A portion of amino acids 50-95 of human SUMO3 (SMT3) was used as the immunogen for this antibody.
Format Purified