Please login to your online account to display your discounted pricing

anti-SUMO3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224651

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224651

  • / Each
 

Description & Specifications

Specifications

Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey and chimp, 93% homologous in cow, dog and horse, 87% homologous in rat and 75% homologous in mouse.
Antigen SUMO3
Format Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. P61957
Host Species Rabbit
Applications Western Blotting
Immunogen A portion of amino acids 50-95 of human SUMO3 (SMT3) was used as the immunogen for this antibody.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6612
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate