Please login to your online account to display your discounted pricing

anti-SUCNR1/GPR91, Polyclonal, Novus Biologicals™

$259.02 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human SUCNR1/GPR91 was used as the immunogen.
Antigen SUCNR1/GPR91
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224549

 
novus biologicals canada
NBP224549
Unlabeled 0.1mg Each for $259.02
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human SUCNR1/GPR91 was used as the immunogen.
Antigen SUCNR1/GPR91
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_c), horse, rat, mouse, cow, dog, rhesus monkey and chimpanzee and 83% homologous in zebrafish.
Gene Accession No. Q9BXA5
Gene ID (Entrez) 56670
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL