Please login to your online account to display your discounted pricing

anti-SUCNR1/GPR91, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224549SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224549SS

  • / Each
 

Description & Specifications

Specifications

Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_c), horse, rat, mouse, cow, dog, rhesus monkey and chimpanzee and 83% homologous in zebrafish.
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Formulation 100μg in 200μL PBS containing 0.05% BSA
Conjugate Unlabeled
Gene Accession No. Q9BXA5
Primary or Secondary Primary
Gene ID (Entrez) 56670
Regulatory Status RUO
Format Affinity Purified
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Primate
Antigen SUCNR1/GPR91
Immunogen A portion of amino acids 100-150 of human SUCNR1/GPR91 was used as the immunogen.
Quantity 0.025mg