Please login to your online account to display your discounted pricing

anti-STRAP, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 230-280 of human STRAP was used as the immunogen.
Antigen STRAP
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224740

 
novus biologicals canada
NBP224740
Unlabeled 0.1mg Each for $255.80

NBP224740SS

 
novus biologicals canada
NBP224740SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 230-280 of human STRAP was used as the immunogen.
Antigen STRAP
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, pig, rat, mouse, chimpanzee, rhesus monkey, orangutan, cow, dog, 88% homologous in xenopus and 83% homologous in drosophila and opossum.
Gene Accession No. Q9Y3F4
Gene ID (Entrez) 11171
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL