Please login to your online account to display your discounted pricing

anti-STK33, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP154156

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP154156

  • / Each
 

Description & Specifications

Specifications

Concentration 0.65mg/mL
Primary or Secondary Primary
Gene Accession No. Q9BYT3
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Gene ID (Entrez) 65975
Formulation PBS, 30% glycerol
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Immunogen A genomic peptide made to an internal region of the human STK33 (within residues 1-150). [Swiss-Prot Q9BYT3]
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Host Species Rabbit
Format Affinity Purified
Quantity 0.1mL
Antigen STK33