Please login to your online account to display your discounted pricing

anti-STK33, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP154156SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP154156SS

  • / Each
 

Description & Specifications

Specifications

Antigen STK33
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.65mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q9BYT3
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human STK33 (within residues 1-150). [Swiss-Prot Q9BYT3]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 65975
Target Species Human
Target Species Murine