Please login to your online account to display your discounted pricing

anti-STING/TMEM173/MPYS, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human STING was used as the immunogen.
Antigen STING/TMEM173/MPYS
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224548

 
novus biologicals canada
NBP224548
Unlabeled 0.1mg Each for $309.05

NBP224548SS

 
novus biologicals canada
NBP224548SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human STING was used as the immunogen.
Antigen STING/TMEM173/MPYS
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), chimpanzee and rhesus monkey, 87% homologous in dog and cow, 81% homologous in horse, mouse and boar and 75% homologous in rat.
Gene Accession No. Q86WV6
Gene ID (Entrez) 340061
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL