Please login to your online account to display your discounted pricing

anti-STING/TMEM173/MPYS, Polyclonal, Novus Biologicals™

$332.66 - $332.66

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the C Terminus Region of the target protein.
Antigen STING/TMEM173/MPYS
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

44320002

 
novus biologicals canada
44320002
Unlabeled 0.05mg Each for $332.66
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the C Terminus Region of the target protein.
Antigen STING/TMEM173/MPYS
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Western Blotting
Test Specificity This product is specific for Human TMEM173/MPYS .
Gene Accession No. Q86WV6
Gene ID (Entrez) 340061
Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0