Please login to your online account to display your discounted pricing

anti-STING/TMEM173/MPYS, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224548SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224548SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Concentration 0.5mg/mL
Primary or Secondary Primary
Gene Accession No. Q86WV6
Formulation 100μg in 200μL PBS containing 0.05% BSA
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), chimpanzee and rhesus monkey, 87% homologous in dog and cow, 81% homologous in horse, mouse and boar and 75% homologous in rat.
Quantity 0.025mg
Antigen STING/TMEM173/MPYS
Gene ID (Entrez) 340061
Immunogen A portion of amino acids 50-100 of human STING was used as the immunogen.
Format Affinity Purified
Host Species Rabbit
Regulatory Status RUO
Cross Reactivity Human
Applications Western Blotting