Please login to your online account to display your discounted pricing

anti-STING/TMEM173/MPYS, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224548

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224548

  • / Each
 

Description & Specifications

Specifications

Antigen STING/TMEM173/MPYS
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. Q86WV6
Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human STING was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 340061
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), chimpanzee and rhesus monkey, 87% homologous in dog and cow, 81% homologous in horse, mouse and boar and 75% homologous in rat.
Target Species Human