Please login to your online account to display your discounted pricing

anti-STIM1, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region (within residues 600-685) of the human Stromal interaction molecule 1 protein. [UniProt No. Q13586]
Antigen STIM1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11060547

 
novus biologicals canada
NB11060547
Unlabeled 0.1mL Each for $309.05

NB11060547T

 
novus biologicals canada
NB11060547SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region (within residues 600-685) of the human Stromal interaction molecule 1 protein. [UniProt No. Q13586]
Antigen STIM1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity Human, mouse, rat, bovine and canine. Immunogen has 93% homology to porcine and 79% homology to Xenopus.
Gene Accession No. Q13586
Gene ID (Entrez) 6786
Formulation PBS, 30% glycerol
Concentration 1mg/mL