Please login to your online account to display your discounted pricing

STB5L Polyclonal Antibody, Invitrogen™


STB5L Antibody, OSS00049W, from Invitrogen™. Species Reactivity: Mouse, Rat; Applications: Immunohistochemistry, Western Blot.

Manufacturer: pierce biotechnology inc  OSS00049W

Catalog No. PIOSS00049W

  • / Each
 

Description & Specifications

Specifications

Antigen STB5L
Applications Immunohistochemistry
Applications Western Blot
Conjugate Unlabeled
Format Serum
Formulation 100μL unpurified antibody in whole serum and no preservative
Gene Accession No. Q5DQR4
Gene Alias Tomosyn-2, Lethal(2) giant larvae protein homolog 4, Stxbp5l, Llgl4, LLGL4, STB5L, MGC169507, 9430094L02, A830015P08Rik
Gene Symbols STXBP5L
Host Species Rabbit
Immunogen A synthetic peptide Rabbit antibody to c-terminal region of mouse Syntaxin-binding protein 5-like
Quantity 100μL
Storage Requirements avoiding freeze/thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 207227, 288080
Target Species Murine
Target Species Rat

STB5L Antibody, OSS00049W, from Invitrogen. Species Reactivity: Mouse, Rat; Applications: Immunohistochemistry, Western Blot.


Host Species: Rabbit
Cross Reactivity: Mouse, Rat
Immunogen: A synthetic peptide Rabbit antibody to c-terminal region of mouse Syntaxin-binding protein 5-like
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: Tomosyn-2, Lethal(2) giant larvae protein homolog 4, Stxbp5l, Llgl4, LLGL4, STB5L, MGC169507, 9430094L02, A830015P08Rik
Uniprot Accession: Q5DQR4
Entrez GeneID: 207227, 288080
Gene Symbol: STXBP5L
Reconstitute with 100 ul of distilled water.