Please login to your online account to display your discounted pricing

anti-STB5L Polyclonal, Thermo Scientific™

STB5L Polyclonal antibody OSS00049W from Thermo Scientific™ specifically detects STB5L in Mouse and Rat samples, and it is validated for Immunohistochemistry and Western Blot

$424.20 - $424.20

Specifications

Antigen STB5L
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Rabbit
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIOSS00050G

 
pierce biotechnology inc
OSS00050G
Unlabeled 500μg Each for $424.20

PIOSS00049W

 
pierce biotechnology inc
OSS00049W
Unlabeled 100μL Each for $424.20
Description & Specifications

Specifications

Antigen STB5L
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Rabbit
Immunogen A synthetic peptide from c-terminal region of mouse Syntaxin-binding protein 5-like
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

STB5L Polyclonal antibody OSS00049W from Thermo Scientific™ specifically detects STB5L in Mouse and Rat samples, and it is validated for Immunohistochemistry and Western Blot


Host Species: Rabbit
Cross Reactivity: Mouse, Rat
Immunogen: A synthetic peptide Rabbit antibody to c-terminal region of mouse Syntaxin-binding protein 5-like
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Tomosyn-2, Lethal(2) giant larvae protein homolog 4, Stxbp5l, Llgl4, LLGL4, STB5L, MGC169507, 9430094L02, A830015P08Rik
Uniprot Accession: Q5DQR4
Gene Symbol: Stxbp5l
Entrez GeneID: 207227, 288080
OSS00049W detects STB5L in Mouse and Rat samples. OSS00049W has been successfully used in Immunohistochemistry and Western Blot procedures. OSS00049W The immunogen is A synthetic peptide Rabbit antibody to c-terminal region of mouse Syntaxin-binding protein 5-like.