Please login to your online account to display your discounted pricing

anti-Stathmin-2/STMN2, Polyclonal, Novus Biologicals™

$139.00 - $384.00

Specifications

Host Species Rabbit
Immunogen C-terminal peptide of mouse STMN2. [Swiss-Prot P55821]
Antigen Stathmin-2/STMN2
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP149461

 
novus biologicals canada
NBP149461
Unlabeled 0.05mL Each for $384.00

NBP149461SS

 
novus biologicals canada
NBP149461SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen C-terminal peptide of mouse STMN2. [Swiss-Prot P55821]
Antigen Stathmin-2/STMN2
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Gene ID (Entrez) 11075