Please login to your online account to display your discounted pricing

anti-Stathmin-2/STMN2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP149461SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP149461SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Immunogen C-terminal peptide of mouse STMN2. [Swiss-Prot P55821]
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 11075
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Stathmin-2/STMN2
Format Antiserum
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Quantity 0.025mL
Regulatory Status RUO