Please login to your online account to display your discounted pricing

anti-Stathmin 1, Polyclonal, Novus Biologicals™

$144.63 - $367.12

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an C-terminal portion of the human Stathmin 1 protein (between residues 100-149) [UniProt P16949]
Antigen Stathmin 1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP176798

 
novus biologicals canada
NBP176798
Unlabeled 0.1mL Each for $367.12

NBP176798SS

 
novus biologicals canada
NBP176798SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an C-terminal portion of the human Stathmin 1 protein (between residues 100-149) [UniProt P16949]
Antigen Stathmin 1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Human, mouse and rat. Predicted to react in bovine, primate, pig, Chinese Hamster and canine based on 100% sequence identity.
Gene Accession No. P16949
Gene ID (Entrez) 3925
Formulation PBS
Concentration 1mg/mL