Please login to your online account to display your discounted pricing

anti-STAT6, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 808-840 of human STAT6. STAT6 antibody will cross-react with mouse and rat
Antigen STAT6
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224826

 
novus biologicals canada
NBP224826
Unlabeled 0.1mg Each for $257.41

NBP224826SS

 
novus biologicals canada
NBP224826SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 808-840 of human STAT6. STAT6 antibody will cross-react with mouse and rat
Antigen STAT6
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene ID (Entrez) 6778
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL