Please login to your online account to display your discounted pricing

anti-STAT6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224826SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224826SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 6778
Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 808-840 of human STAT6. STAT6 antibody will cross-react with mouse and rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Antigen STAT6
Quantity 0.025mg
Isotype IgG
Format Purified