Please login to your online account to display your discounted pricing

anti-STAT6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224826

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224826

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6778
Concentration 0.5mg/mL
Conjugate Unlabeled
Antigen STAT6
Format Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 808-840 of human STAT6. STAT6 antibody will cross-react with mouse and rat
Isotype IgG
Quantity 0.1mg
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Host Species Rabbit
Regulatory Status RUO