missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-STAT5b, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This polyclonal antibody was developed against a synthetic peptide corresponding to amino acids 771-787 of human Stat5b.
Antigen STAT5b
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056324T Novus Biologicals Canada
NB10056324SS
0.025mg Each for $139.00
NB10056324 Novus Biologicals Canada
NB10056324
0.1mg Each for $255.80
Specifications Description & Specifications

Specifications

Rabbit
STAT5b
Purified
RUO
In Jurkat, a 92 kDa band is observed. Cross reacts with Human. Predicted to react with Bovine, Chicken, Mouse, New World Monkey and Rat.
6777
0.5mg/mL
This polyclonal antibody was developed against a synthetic peptide corresponding to amino acids 771-787 of human Stat5b.
Polyclonal
Primary
Western Blot
P51692
PBS containing 0.05% BSA