Please login to your online account to display your discounted pricing

anti-STAT5b, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056324SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056324T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P51692
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Antigen STAT5b
Immunogen This polyclonal antibody was developed against a synthetic peptide corresponding to amino acids 771-787 of human Stat5b.
Primary or Secondary Primary
Test Specificity In Jurkat, a 92 kDa band is observed. Cross reacts with Human. Predicted to react with Bovine, Chicken, Mouse, New World Monkey and Rat.
Concentration 0.5mg/mL
Cross Reactivity Human
Formulation PBS containing 0.05% BSA
Host Species Rabbit
Quantity 0.025mg
Format Purified
Applications Western Blotting
Regulatory Status RUO
Gene ID (Entrez) 6777