Please login to your online account to display your discounted pricing

anti-STAT2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056323

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056323

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Primary or Secondary Primary
Gene Accession No. NP_005410.1
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6773
Test Specificity In Jurkat, an approx. 113 kDa band is observed.
Formulation PBS containing 0.05% BSA
Immunogen This polyclonal antibody was developed against a synthetic peptide corresponding to amino acids 830-849 of human Stat2.
Regulatory Status RUO
Host Species Rabbit
Antigen STAT2
Applications Western Blotting
Cross Reactivity Human
Format Purified
Quantity 0.1mg