Please login to your online account to display your discounted pricing

anti-STAT2, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This polyclonal antibody was developed against a synthetic peptide corresponding to amino acids 830-849 of human Stat2.
Antigen STAT2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056323

 
novus biologicals canada
NB10056323
Unlabeled 0.1mg Each for $255.80

NB10056323T

 
novus biologicals canada
NB10056323SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This polyclonal antibody was developed against a synthetic peptide corresponding to amino acids 830-849 of human Stat2.
Antigen STAT2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity In Jurkat, an approx. 113 kDa band is observed.
Gene Accession No. NP_005410.1
Gene ID (Entrez) 6773
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL