Please login to your online account to display your discounted pricing

anti-STAT2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056323SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056323T

  • / Each
 

Description & Specifications

Specifications

Antigen STAT2
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. NP_005410.1
Immunogen This polyclonal antibody was developed against a synthetic peptide corresponding to amino acids 830-849 of human Stat2.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6773
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Host Species Rabbit
Applications Western Blotting
Quantity 0.025mg
Regulatory Status RUO
Test Specificity In Jurkat, an approx. 113 kDa band is observed.