Please login to your online account to display your discounted pricing

anti-STAT1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide sequence corresponding to amino acids between 700 and 750 of human Stat1 was used as immunogen. This sequence is 100% conserved between Stat1 alpha and Stat1 beta.
Antigen STAT1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056314

 
novus biologicals canada
NB10056314
Unlabeled 0.1mg Each for $255.80

NB10056314T

 
novus biologicals canada
NB10056314SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide sequence corresponding to amino acids between 700 and 750 of human Stat1 was used as immunogen. This sequence is 100% conserved between Stat1 alpha and Stat1 beta.
Antigen STAT1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity This antibody is able to detect both the alpha form (p91, ∽91 kD) and the beta form (p84, ∽84 kD) of STAT1. Reacts with Chimpanzee, Canine, Human, Mouse and Porcine. Predicted to react with Rat (86%) and Chicken (80%).
Gene Accession No. NP_009330.1
Gene ID (Entrez) 6772
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL