Please login to your online account to display your discounted pricing

anti-ST2/IL-1 R4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224736SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224736SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6761
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_d) and 94% homologous in chimpanzee.
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Purified
Regulatory Status RUO
Antigen ST2/IL-1 R4
Formulation 100μg in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen A portion of amino acids 25-65 of human ST2 protein was used as the immunogen.
Isotype IgG
Cross Reactivity Human
Quantity 0.025mg