Please login to your online account to display your discounted pricing

anti-SSEA-4, NL557, Clone: MC-813-70, R&D Systems


Manufacturer: r & d systems  RDSNLLC1435R

 View more versions of this product

Catalog No. NLLC1435R

  • / Each
 

Description & Specifications

Specifications

Antigen SSEA-4
Quantity 0.5mL
Cross Reactivity Human
Cross Reactivity Mouse
Regulatory Status RUO
Immunogen 2120Ep human embryonal carcinoma cell line.
Clone MC-813-70
Conjugate NL557
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Purification Method Protein A or G purified from hybridoma culture supernatant
Isotype IgG3
Format Purified