Please login to your online account to display your discounted pricing

anti-SSEA-4, NL557, Clone: MC-813-70, R&D Systems

$452.99 - $585.69

Specifications

Antigen SSEA-4
Clone MC-813-70
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NLLC1435R

 
r & d systems
RDSNLLC1435R
NL557 0.5mL Each for $585.69

NL1435R

 
r & d systems
RDSNL1435R
NL557 0.5mL Each for $452.99
Description & Specifications

Specifications

Antigen SSEA-4
Clone MC-813-70
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Host Species Murine
Regulatory Status RUO
Immunogen 2120Ep human embryonal carcinoma cell line.
Isotype IgG3
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Immunocytochemistry