Please login to your online account to display your discounted pricing

anti-SSEA-4, NL493, Clone: MC-813-70, R&D Systems


Manufacturer: r & d systems  RDSNLLC1435G

 View more versions of this product

Catalog No. NLLC1435G

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Quantity 0.5mL
Regulatory Status RUO
Host Species Mouse
Antigen SSEA-4
Isotype IgG3
Clone MC-813-70
Conjugate NL493
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Format Purified
Immunogen 2120Ep human embryonal carcinoma cell line.
Purification Method Protein A or G purified from hybridoma culture supernatant